Cobalt(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Cobalt(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 035730.06
also known as 035730-06
Price (USD)/ Each
454.65
Online exclusive
505.00 
Save 50.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
454.65
Online exclusive
505.00 
Save 50.35 (10%)
Add to cart
Cobalt(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number035730.06
Price (USD)/ Each
454.65
Online exclusive
505.00 
Save 50.35 (10%)
-
Add to cart
Chemical Identifiers
CAS15238-00-3
IUPAC Nameλ²-cobalt(2+) diiodide
Molecular FormulaCoI2
InChI KeyAVWLPUQJODERGA-UHFFFAOYSA-L
SMILES[Co++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.001% max.
Particle Size-10 Mesh
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only