Gadolinium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Gadolinium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gadolinium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 035735.03
also known as 035735-03
Price (USD)/ Each
136.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
136.00
Add to cart
Gadolinium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Catalog number035735.03
Price (USD)/ Each
136.00
-
Add to cart
Chemical Identifiers
CAS13572-98-0
IUPAC Namegadolinium(3+) triiodide
Molecular FormulaGdI3
InChI KeyIZZTUGMCLUGNPM-UHFFFAOYSA-K
SMILES[I-].[I-].[I-].[Gd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Rare Earth Oxide Impurities0.01% max.
Appearance (Color)Yellow
Residual water< 100 ppm
It is used as a compound in semiconductors and other high purity applications, for contrast imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only