Thermo Scientific Chemicals

Gadolinium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 035736.06
5 g, Each
Thermo Scientific Chemicals

Gadolinium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 035736.06
5 g, Each
Quantity
Catalog number: 035736.06
also known as 035736-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10138-52-0
IUPAC Name
gadolinium(3+) trichloride
Molecular Formula
Cl3Gd
InChI Key
MEANOSLIBWSCIT-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Gd+3]
Appearance (Color)
White
Residual water
< 100 ppm
Form
Powder
Total Rare Earth Oxide Impurities
0.01% max.

Description

Gadolinium(III) Chloride is used as chelating agents, which is essential for biomedical applications. It is also employed in the synthesis and nuclear magnetic resonance (NMR) studies of paramagnetic GdPCTA-[12] and GdPCTA-[13] complexes. It plays an important role as contrast agent for magnetic resonance imaging and an macrophage inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text