Gadolinium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Gadolinium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gadolinium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 035736.14
also known as 035736-14
Price (USD)/ Each
594.65
Online exclusive
660.00 
Save 65.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
594.65
Online exclusive
660.00 
Save 65.35 (10%)
Add to cart
Gadolinium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Catalog number035736.14
Price (USD)/ Each
594.65
Online exclusive
660.00 
Save 65.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10138-52-0
IUPAC Namegadolinium(3+) trichloride
Molecular FormulaCl3Gd
InChI KeyMEANOSLIBWSCIT-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Gd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Residual water< 100 ppm
FormPowder
Total Rare Earth Oxide Impurities0.01% max.
Gadolinium(III) Chloride is used as chelating agents, which is essential for biomedical applications. It is also employed in the synthesis and nuclear magnetic resonance (NMR) studies of paramagnetic GdPCTA-[12] and GdPCTA-[13] complexes. It plays an important role as contrast agent for magnetic resonance imaging and an macrophage inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only