Thermo Scientific Chemicals

Europium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 035738.06
5 g, Each
Thermo Scientific Chemicals

Europium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 035738.06
5 g, Each
Quantity
Catalog number: 035738.06
also known as 035738-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-76-0
IUPAC Name
europium(3+) trichloride
Molecular Formula
Cl3Eu
InChI Key
NNMXSTWQJRPBJZ-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Eu+3]
Total Rare Earth Oxide Impurities
0.01% max.
Appearance (Color)
Yellow
Residual water
< 100 ppm
Form
Powder

Description

Europium(III) chloride is used as a starting material for the preparation of other europium salts. It is also used in preparation of europium(II)chloride by reduction in a gold boat using hydrogen gas. Further, it is also used to prepare organometallic compounds of europium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text