Europium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Europium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Europium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number 035738.14
also known as 035738-14
Price (USD)/ Each
945.65
Online exclusive
1051.00 
Save 105.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
945.65
Online exclusive
1051.00 
Save 105.35 (10%)
Add to cart
Europium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Catalog number035738.14
Price (USD)/ Each
945.65
Online exclusive
1051.00 
Save 105.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10025-76-0
IUPAC Nameeuropium(3+) trichloride
Molecular FormulaCl3Eu
InChI KeyNNMXSTWQJRPBJZ-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Eu+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Rare Earth Oxide Impurities0.01% max.
Appearance (Color)Yellow
Residual water< 100 ppm
Europium(III) chloride is used as a starting material for the preparation of other europium salts. It is also used in preparation of europium(II)chloride by reduction in a gold boat using hydrogen gas. Further, it is also used to prepare organometallic compounds of europium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only