Thermo Scientific Chemicals

Erbium(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035739.14
25 g, Each
Thermo Scientific Chemicals

Erbium(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035739.14
25 g, Each
Quantity
Catalog number: 035739.14
also known as 035739-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10138-41-7
IUPAC Name
erbium(3+) trichloride
Molecular Formula
Cl3Er
InChI Key
HDGGAKOVUDZYES-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Er+3]
Residual water
< 100 ppm
Appearance (Color)
Pink
Form
Color
Total Rare Earth Oxide Impurities
0.1% max.

Description

It is a starting material for new edge-bridged octahedral M6 cluster compounds which are used for electron and stability studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text