Thermo Scientific Chemicals

Yttrium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 035743.18
50 g, Each
Thermo Scientific Chemicals

Yttrium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 035743.18
50 g, Each
Quantity
Catalog number: 035743.18
also known as 035743-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10361-92-9
IUPAC Name
yttrium(3+) hexahydrate trichloride
Molecular Formula
Cl3H12O6Y
InChI Key
IINACGXCEZNYTF-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Y+3]
Appearance (Color)
White
Residual water
< 100 ppm
Form
Powder
Total Rare Earth Oxide Impurities
0.01% max.

Description

Yttrium(III) chloride is used as a precursor for titanium-containing ceramics. It is used as an analytical reagent, as a catalyst and in superconductors. It is also used in the production of yttrium alkyl alkoxide complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text