Yttrium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Yttrium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number 035743.30
also known as 035743-30
Price (USD)/ Each
1,055.65
Online exclusive
1173.00 
Save 117.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
1,055.65
Online exclusive
1173.00 
Save 117.35 (10%)
Add to cart
Yttrium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Catalog number035743.30
Price (USD)/ Each
1,055.65
Online exclusive
1173.00 
Save 117.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10361-92-9
IUPAC Nameyttrium(3+) hexahydrate trichloride
Molecular FormulaCl3H12O6Y
InChI KeyIINACGXCEZNYTF-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Residual water< 100 ppm
FormPowder
Total Rare Earth Oxide Impurities0.01% max.
Yttrium(III) chloride is used as a precursor for titanium-containing ceramics. It is used as an analytical reagent, as a catalyst and in superconductors. It is also used in the production of yttrium alkyl alkoxide complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only