Iridium, plasma standard solution, Specpure™ Ir 10,000μg/mL, Thermo Scientific Chemicals
Iridium, plasma standard solution, Specpure™ Ir 10,000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iridium, plasma standard solution, Specpure™ Ir 10,000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
50 mL
100 mL
Catalog number 035752.AP
also known as 035752-AP
Price (USD)/ Each
-
Quantity:
500 mL
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialIridium
CAS7439-88-5
Health Hazard 1H290-H314-H335
Quantity500 mL
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only