Europium, plasma standard solution, Specpure™ Eu 1000μg/mL, Thermo Scientific Chemicals
Europium, plasma standard solution, Specpure™ Eu 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Europium, plasma standard solution, Specpure™ Eu 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 035753.AD
also known as 035753-AD
Price (USD)/ Each
51.70
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
51.70
Add to cart
Europium, plasma standard solution, Specpure™ Eu 1000μg/mL, Thermo Scientific Chemicals
Catalog number035753.AD
Price (USD)/ Each
51.70
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialEuropium plasma standard solution
ConcentrationEu2O3 in 5% HNO3
Name NoteEu 1000μg/mL
Health Hazard 1H290-H314-H335
Quantity50 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only