Rhodium, plasma standard solution, Specpure™ Rh 1000μg/mL, Thermo Scientific Chemicals
Rhodium, plasma standard solution, Specpure™ Rh 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhodium, plasma standard solution, Specpure™ Rh 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
50 mL
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035754.AP
also known as 035754-AP
Price (USD)/ Each
1,190.00
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
1,190.00
Add to cart
Rhodium, plasma standard solution, Specpure™ Rh 1000μg/mL, Thermo Scientific Chemicals
Catalog number035754.AP
Price (USD)/ Each
1,190.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialRhodium
Name NoteRh 1000μg/mL
CAS7440-16-6
Health Hazard 1H290-H314-H335
Quantity500 mL
View more
Rhodium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only