Rhodium, plasma standard solution, Specpure™ Rh 10,000μg/mL, Thermo Scientific Chemicals
Rhodium, plasma standard solution, Specpure™ Rh 10,000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhodium, plasma standard solution, Specpure™ Rh 10,000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
50 mL
Catalog number 035763.AE
also known as 035763-AE
Price (USD)/ Each
-
Quantity:
100 mL
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialRhodium plasma standard solution
ConcentrationRhCl3 in 20% HCl
Name NoteRh 10000μg/mL
Health Hazard 1H290-H314-H331-H335-H341
Quantity100 mL
View more
Rhodium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only