Thermo Scientific Chemicals

Zinc chloride hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 035782.30
250 g, Each
Thermo Scientific Chemicals

Zinc chloride hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 035782.30
250 g, Each
Quantity
Catalog number: 035782.30
also known as 035782-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29604-34-0
IUPAC Name
zinc(2+) dichloride
Molecular Formula
Cl2Zn
InChI Key
JIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Zn++]
Total Metal Impurities
0.001% max.

Description

It is used for a source of the zinc ion. It can also be used alone or with phenols for preserving railway ties, fireproofing timber, and with ammonium chloride as a flux for soldering and etching of metals. It is used as a moderate-strength lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text