Zinc acetate, anhydrous, 99.98% (metals basis), Thermo Scientific Chemicals
Zinc acetate, anhydrous, 99.98% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc acetate, anhydrous, 99.98% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 035792.09
also known as 035792-09
Price (USD)/ Each
76.65
Online exclusive
85.00 
Save 8.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
76.65
Online exclusive
85.00 
Save 8.35 (10%)
Add to cart
Zinc acetate, anhydrous, 99.98% (metals basis), Thermo Scientific Chemicals
Catalog number035792.09
Price (USD)/ Each
76.65
Online exclusive
85.00 
Save 8.35 (10%)
-
Add to cart
Chemical Identifiers
CAS557-34-6
IUPAC Namezinc(2+) diacetate
Molecular FormulaC4H6O4Zn
InChI KeyDJWUNCQRNNEAKC-UHFFFAOYSA-L
SMILES[Zn++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.02% max.
Zinc acetate anhydrous is used in the synthesis of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis. It is utilized in the ultrasonic preparation of zinc sulfide nanoparticles coated on silica particles. It is administered orally or parenterally as a nutritional supplement. It finds an application in the field of industries such as wood preservation, manufacturing other zinc salts, polymers, manufacture of ethylene acetate, as a dye mordant, and analytical reagent. It also acts as a plating inhibitor on primary water piping.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only