Thermo Scientific Chemicals

Zinc acetate, anhydrous, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 035792.18
50 g, Each
Thermo Scientific Chemicals

Zinc acetate, anhydrous, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 035792.18
50 g, Each
Quantity
Catalog number: 035792.18
also known as 035792-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
557-34-6
IUPAC Name
zinc(2+) diacetate
Molecular Formula
C4H6O4Zn
InChI Key
DJWUNCQRNNEAKC-UHFFFAOYSA-L
SMILES
[Zn++].CC([O-])=O.CC([O-])=O
Total Metal Impurities
0.02% max.

Description

Zinc acetate anhydrous is used in the synthesis of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis. It is utilized in the ultrasonic preparation of zinc sulfide nanoparticles coated on silica particles. It is administered orally or parenterally as a nutritional supplement. It finds an application in the field of industries such as wood preservation, manufacturing other zinc salts, polymers, manufacture of ethylene acetate, as a dye mordant, and analytical reagent. It also acts as a plating inhibitor on primary water piping.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text