Strontium carbonate, 99.99% (metals basis), Thermo Scientific Chemicals
Strontium carbonate, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontium carbonate, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 035793.36
also known as 035793-36
Price (USD)/ Each
1,263.65
Online exclusive
1404.00 
Save 140.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
1,263.65
Online exclusive
1404.00 
Save 140.35 (10%)
Add to cart
Strontium carbonate, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number035793.36
Price (USD)/ Each
1,263.65
Online exclusive
1404.00 
Save 140.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1633-05-2
IUPAC Namestrontium(2+) carbonate
Molecular FormulaCO3Sr
InChI KeyLEDMRZGFZIAGGB-UHFFFAOYSA-L
SMILES[Sr++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Used as a colorant in fireworks. It is also used in ceramics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only