Niobium(V) chloride, 99.95% (metals basis), Thermo Scientific Chemicals
Niobium(V) chloride, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium(V) chloride, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 035795.09
also known as 035795-09
Price (USD)/ Each
86.65
Online exclusive
96.00 
Save 9.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
86.65
Online exclusive
96.00 
Save 9.35 (10%)
Add to cart
Niobium(V) chloride, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number035795.09
Price (USD)/ Each
86.65
Online exclusive
96.00 
Save 9.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10026-12-7
IUPAC Nameniobium(5+) pentachloride
Molecular FormulaCl5Nb
InChI KeyYHBDIEWMOMLKOO-UHFFFAOYSA-I
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Nb+5]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow white
FormCrystalline
Total Metal Impurities0.05% max.
It is used to form mixed chloro-aryloxide compounds, such as [Nb(OC6H3-2,6-i-Pr2)2Cl3]2, which form pyridine or phosphine adducts with a distorted six-coordinate geometry. It is also used in the preparation of pure niobium, and as an intermediate, precursor to many other laboratory reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only