Thermo Scientific Chemicals

Lanthanum(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035800.04
2 g, Each
Thermo Scientific Chemicals

Lanthanum(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035800.04
2 g, Each
Quantity
Catalog number: 035800.04
also known as 035800-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13536-79-3
IUPAC Name
lanthanum(3+) tribromide
Molecular Formula
Br3La
InChI Key
XKUYOJZZLGFZTC-UHFFFAOYSA-K
SMILES
[Br-].[Br-].[Br-].[La+3]
Appearance (Color)
White
Residual water
< 100 ppm
Total Rare Earth Oxide Impurities
0.1% max.
Particle Size
-10 Mesh
Form
Beads

Description

It is used as a scintillation material in certain application, as a source in the synthesis of lanthanum & its derivatives CsNaLa6Br14N2 and La3Br6N which contain rare bitetrahedral structural units..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text