Thermo Scientific Chemicals

Lanthanum(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035801.06
5 g, Each
Thermo Scientific Chemicals

Lanthanum(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035801.06
5 g, Each
Quantity
Catalog number: 035801.06
also known as 035801-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13813-22-4
IUPAC Name
lanthanum(3+) triiodide
Molecular Formula
I3La
InChI Key
KYKBXWMMXCGRBA-UHFFFAOYSA-K
SMILES
[I-].[I-].[I-].[La+3]
Appearance (Color)
White
Form
Beads
Residual water
< 100 ppm
Total Rare Earth Oxide Impurities
0.1% max.
Particle Size
-10 Mesh

Description

Lanthanum(III) iodide is used as a catalyst in the transesterification of N-acyloxazolidinones with alcohol gives the corresponding esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text