Lutetium(III) chloride, ultra dry, 99.98% (REO), Thermo Scientific Chemicals
Lutetium(III) chloride, ultra dry, 99.98% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lutetium(III) chloride, ultra dry, 99.98% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 035802.06
also known as 035802-06
Price (USD)/ Each
513.65
Online exclusive
570.00 
Save 56.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
513.65
Online exclusive
570.00 
Save 56.35 (10%)
Add to cart
Lutetium(III) chloride, ultra dry, 99.98% (REO), Thermo Scientific Chemicals
Catalog number035802.06
Price (USD)/ Each
513.65
Online exclusive
570.00 
Save 56.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10099-66-8
IUPAC Namelutetium(3+) trichloride
Molecular FormulaCl3Lu
InChI KeyAEDROEGYZIARPU-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Lu+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water< 100 ppm
Total Rare Earth Oxide Impurities0.02% max.
Appearance (Color)White
FormPowder
It is used as an additive for chemical industry, single crystal scintillators, carbon arc rods, PET scan detectors, high refractive index glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only