Zirconium carbide, 99.5% (metals basis excluding Hf), Hf< 200ppm, Thermo Scientific Chemicals
Zirconium carbide, 99.5% (metals basis excluding Hf), Hf&lt; 200ppm, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium carbide, 99.5% (metals basis excluding Hf), Hf< 200ppm, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035808.18
Price (USD)/ Each
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS12070-14-3
IUPAC Namemethanidylidynezirconiumylium
Molecular FormulaCZr
InChI KeyMHMXYYDKIQTOHA-UHFFFAOYSA-N
SMILES[C-]#[Zr+]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentHf<200ppm
Total Metal Impurities excluding Hf0.5% max.
Zirconium carbide commercially used in tool bits for cutting tools. It is used as an abrasive, in cladding, in cermets and incandescent filaments. It finds application in the coating of uranium dioxide and thorium dioxide particles of nuclear fuel as well as in thermo-photovoltaic radiators and field emitter tips and arrays. It is involved in the usage as refractory coatings in nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only