Thermo Scientific Chemicals

Zirconium carbide, 99.5% (metals basis excluding Hf), Hf< 200ppm, Thermo Scientific Chemicals

Catalog number: 035808.30
250 g, Each
Thermo Scientific Chemicals

Zirconium carbide, 99.5% (metals basis excluding Hf), Hf< 200ppm, Thermo Scientific Chemicals

Catalog number: 035808.30
250 g, Each
Quantity
Catalog number: 035808.30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12070-14-3
IUPAC Name
methanidylidynezirconiumylium
Molecular Formula
CZr
InChI Key
MHMXYYDKIQTOHA-UHFFFAOYSA-N
SMILES
[C-]#[Zr+]
Impurity content
Hf<200ppm
Total Metal Impurities excluding Hf
0.5% max.

Description

Zirconium carbide commercially used in tool bits for cutting tools. It is used as an abrasive, in cladding, in cermets and incandescent filaments. It finds application in the coating of uranium dioxide and thorium dioxide particles of nuclear fuel as well as in thermo-photovoltaic radiators and field emitter tips and arrays. It is involved in the usage as refractory coatings in nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text