Titanium Thinfoil, 0.006mm (0.0002in) thick, 99.6% (metals basis), Thermo Scientific Chemicals
Titanium Thinfoil, 0.006mm (0.0002in) thick, 99.6% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium Thinfoil, 0.006mm (0.0002in) thick, 99.6% (metals basis), Thermo Scientific Chemicals

Form: Approx. 0.02g/25 x 25mm; Not certified pinhole free
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number 035813.FI
also known as 035813-FI
Price (USD)/ Each
623.00
-
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)/ Each
623.00
Add to cart
Titanium Thinfoil, 0.006mm (0.0002in) thick, 99.6% (metals basis), Thermo Scientific Chemicals
Catalog number035813.FI
Price (USD)/ Each
623.00
-
Add to cart
Specifications
Chemical Name or MaterialTitanium
ColorSilver to Gray
CAS7440-32-6
AppearanceSilver Gray foil
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only