Specifications
Chemical Name or MaterialNickel Thinfoil
Name Note0.0125mm (0.000492 in.) thick, Not certified pinhole free
CAS7440-02-0
Health Hazard 1H317-H351-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
View more