Thermo Scientific Chemicals

Bismuth(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 035834.30
250 g, Each
Thermo Scientific Chemicals

Bismuth(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 035834.30
250 g, Each
Quantity
Catalog number: 035834.30
also known as 035834-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1345-07-9
IUPAC Name
dibismuth(3+) trisulfanediide
Molecular Formula
Bi2S3
InChI Key
YNRGZHRFBQOYPP-UHFFFAOYSA-N
SMILES
[S--].[S--].[S--].[Bi+3].[Bi+3]
Total Metal Impurities
0.1% max.

Description

Bismuth(III) sulfide is used as a starting material to produce other bismuth compounds. It is used in glass industry as a colorant, electronic ceramics, scintillator BGO and in superconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text