Thermo Scientific Chemicals

Cerium silicide, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035836.14
25 g, Each
Thermo Scientific Chemicals

Cerium silicide, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 035836.14
25 g, Each
Quantity
Catalog number: 035836.14
also known as 035836-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12014-85-6
IUPAC Name
(silanylidenecerio)silane
Molecular Formula
CeH4Si2
InChI Key
YARAWHILSWOVAK-UHFFFAOYSA-N
SMILES
[SiH2]=[Ce]=[SiH2]
Appearance (Color)
Grey
Form
Pieces, 3-15mm
Total Rare Earth Oxide Impurities
≤0.1%

Description

Cerium silicide is used as a surrogate material to investigate the feasibility of high-energy ball milling and Spark Plasma Sintering (SPS) for the production of uranium silicide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text