Thermo Scientific Chemicals

Boron tribromide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 035841.09
Each
Thermo Scientific Chemicals

Boron tribromide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 035841.09
Each
Catalog number: 035841.09
also known as 035841-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10294-33-4
IUPAC Name
tribromoborane
Molecular Formula
BBr3
InChI Key
ILAHWRKJUDSMFH-UHFFFAOYSA-N
SMILES
BrB(Br)Br
Total Metal Impurities
0.01% max.

Description

Boron tribromide, is used commonly in Pharmaceutical manufacturing, image processing, semiconductor doping, semiconductor plasma etching, photovoltaic manufacturing, reagent for various chemical processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text