Thermo Scientific Chemicals

Iron arsenide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 036190.09
10 g, Each
Thermo Scientific Chemicals

Iron arsenide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 036190.09
10 g, Each
Quantity
Catalog number: 036190.09
also known as 036190-09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12005-88-8
IUPAC Name
ferrio-λ²-arsanyl iron
Molecular Formula
AsFe2
InChI Key
FDKAYGUKROYPRO-UHFFFAOYSA-N
SMILES
[Fe].[Fe]=[As]
Total Metal Impurities
0.5% max.

Description

Iron arsenide is used as a superconductor, semiconductor and in photo optic applications 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text