Thermo Scientific Chemicals

Lithium sulfate monohydrate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 036216.A3
2 kg, Each
Thermo Scientific Chemicals

Lithium sulfate monohydrate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 036216.A3
2 kg, Each
Quantity
Catalog number: 036216.A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10102-25-7
IUPAC Name
dilithium(1+) hydrate sulfate
Molecular Formula
H2Li2O5S
InChI Key
RKGLUDFWIKNKMX-UHFFFAOYSA-L
SMILES
[Li+].[Li+].O.[O-]S([O-])(=O)=O
Assay (unspecified)
99.0% Li₂SO₄ min
Impurity content
Cl 0.002%
Impurity content
Fe 0.001%
Impurity content
K 0.05%
Impurity content
Na 0.05%

Description

Lithium sulfate monohydrate is used in solar panels and potential for a new class of battery. It is also used to treat bipolar disorder and in ultrasound-type non-destructive testing. It is also used as an additive for portland cement to speed up the curing. It is also involved as a catalyst in the elimination reaction for the conversion of n-butyl bromide to 1-butene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text