Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals

α-D-Lactose monohydrate, ACS

Catalog number: 036218.A3
2 kg, Each
Thermo Scientific Chemicals

α-D-Lactose monohydrate, ACS

Catalog number: 036218.A3
2 kg, Each
Quantity
Catalog number: 036218.A3
also known as 036218-A3
Price (USD)
Have Questions?

Chemical Identifiers

CAS
64044-51-5
IUPAC Name
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
Molecular Formula
C12H24O12
InChI Key
WSVLPVUVIUVCRA-RJMJUYIDSA-N
SMILES
O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
Heavy Metals (as Pb)
5ppm, max
Appearance (Color)
White
Form
powder
Residue on ignition
0.03%, max
Impurity content
Dextrose P.T.

Description

It is also heavily used in the pharmaceutical industry as a pill filler. They are capable of fermenting lactose for ethanol production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• μa-D-Lactose monohydrate, ACS is the crystalline form of D-Lactose
• It is composed of galactose and glucose subunits

Application

• μa-D-Lactose monohydrate can be used to ferment lactose for ethanol production in laboratory settings
• It may be used as a reference standard for lactose quantitation

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text