Thermo Scientific Chemicals

Citric acid diammonium salt, ACS, 98.0-103.0%, Thermo Scientific Chemicals

Catalog number: 036219.A3
2 kg, Each
Thermo Scientific Chemicals

Citric acid diammonium salt, ACS, 98.0-103.0%, Thermo Scientific Chemicals

Catalog number: 036219.A3
2 kg, Each
Quantity
Catalog number: 036219.A3
also known as 036219-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3012-65-5
IUPAC Name
diammonium 3-carboxy-3-hydroxypentanedioate
Molecular Formula
C6H14N2O7
InChI Key
YXVFQADLFFNVDS-UHFFFAOYSA-N
SMILES
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
Impurity content
SO₄ 0.005%
Insoluble material
0.005%
Trace Metals
(as Pb) 5ppm
Residue on ignition
0.01%
Impurity content
C₂O₄ P.T. (limit about 0.05%)

Description

Citric acid diammonium salt is used in pharmaceuticals, rust proofing, cotton printing and plasticizers. It is also used as a reagent for the determination of phosphate in fertilizers, and as a buffering agent, chelating agent and source of nitrogen in chicken food supplements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text