Thermo Scientific Chemicals

Ammonium oxalate monohydrate, ACS, 99.0-101.0%, Thermo Scientific Chemicals

Catalog number: 036228.57
4 x 500 g, Each
Thermo Scientific Chemicals

Ammonium oxalate monohydrate, ACS, 99.0-101.0%, Thermo Scientific Chemicals

Catalog number: 036228.57
4 x 500 g, Each
Quantity
Catalog number: 036228.57
also known as 036228-57
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6009-70-7
IUPAC Name
oxalic acid diamine hydrate
Molecular Formula
C2H10N2O5
InChI Key
MSMNVXKYCPHLLN-UHFFFAOYSA-N
SMILES
N.N.O.OC(=O)C(O)=O
Impurity content
Cl 0.002%
Impurity content
Fe 2ppm
Impurity content
SO₄ 0.002%
Insoluble material
0.005%
Trace Metals
(as Pb) 5ppm

Description

In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metalsAmmonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metals
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text