Benzoic acid, ACS, 99.5% min, Thermo Scientific Chemicals
Benzoic acid, ACS, 99.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzoic acid, ACS, 99.5% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 036230.22
also known as 036230-22
Price (USD)/ Each
44.65
Online exclusive
49.00 
Save 4.35 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
44.65
Online exclusive
49.00 
Save 4.35 (9%)
Add to cart
Benzoic acid, ACS, 99.5% min, Thermo Scientific Chemicals
Catalog number036230.22
Price (USD)/ Each
44.65
Online exclusive
49.00 
Save 4.35 (9%)
-
Add to cart
Chemical Identifiers
CAS65-85-0
IUPAC Namebenzoic acid
Molecular FormulaC7H6O2
InChI KeyWPYMKLBDIGXBTP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition0.005%
Impurity contentChlorine compounds (as Cl) 0.005%
Impurity contentInsoluble in methanol 0.005%
Impurity contentSubstances reducing permanganate P.T.
Impurity contentSulfur compounds (as S) 0.002%
View more
Benzoic acid is an important precursor for the preparation of many other organic substances viz. benzoyl chloride, benylbenzoate, phenol, sodium benzoate, benzyl alcohol as well as benzoate plasticizers such as glycol-, diethhyleneglycol- and triethyleneglycol esters. Benzoic acid and its salts are used as food preservatives. It is involved in rubber polymerization as an activator and a retardant. It is the main component of benzoin resin and is a constituent of Whitfield’s ointment, which is used for the treatment of fungal skin diseases such as tinea, ringworm and athlete’s foot. It is widely used in cosmetics, dyes, plastics and insect repellents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only