L-(+)-Ascorbic acid, ACS, 99+%, Thermo Scientific Chemicals
L-(+)-Ascorbic acid, ACS, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-(+)-Ascorbic acid, ACS, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 036237.14
also known as 036237-14
Price (USD)/ Each
24.65
Online exclusive
26.70 
Save 2.05 (8%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
24.65
Online exclusive
26.70 
Save 2.05 (8%)
Add to cart
L-(+)-Ascorbic acid, ACS, 99+%, Thermo Scientific Chemicals
Catalog number036237.14
Price (USD)/ Each
24.65
Online exclusive
26.70 
Save 2.05 (8%)
-
Add to cart
Chemical Identifiers
CAS50-81-7
IUPAC Name(5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
Molecular FormulaC6H8O6
InChI KeyCIWBSHSKHKDKBQ-DOMZIZNONA-N
SMILESOC[C@H](O)[C@H]1OC(=O)C(O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥99.0%
Heavy Metals≤0.002%
Residue on ignition≤0.1%
Impurity contentFe: ≤0.001%
Optical RotationA25/D +21.0 ±0.5°
It is all important nutrient and antioxidant. L-ascorbic acid is frequently practiced as a processing aid or as a preservative in certain nutrients or food components. It is likewise utilized in the synthetic polymer industry, in photo processing, and in metal technology. Miscellaneous uses have been proposed in cosmetics, fibers, preservation of blood, preservation of cut plants, cleaning agents, and in assay reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only