Yttrium aluminum oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Yttrium aluminum oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium aluminum oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Can be used as a sputtering target to prepare bubble memory devices
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 036241.14
also known as 036241-14
Price (USD)/ Each
121.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
121.00
Add to cart
Yttrium aluminum oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number036241.14
Price (USD)/ Each
121.00
-
Add to cart
Chemical Identifiers
CAS12005-21-9
IUPAC Namealuminium(3+) yttrium(3+) trioxidandiide
Molecular FormulaAlO3Y
InChI KeyJNDMLEXHDPKVFC-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Can be used as a sputtering target to prepare bubble memory devices.Yttrium aluminum oxide finds application in ceramic, glass and optics due to its thermal stability and insolubility. It is used in the preparation of sputtering bonded targets in chemical processes viz. chemical vapor deposition, atomic thermal and electron beam evaporation. It is also used in solar, water treatment and fuel cell. Further, it is used as coloring and adhesive agents in enamels manufacture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only