Ammonium thiocyanate, ACS, 97.5% min, Thermo Scientific Chemicals
Ammonium thiocyanate, ACS, 97.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium thiocyanate, ACS, 97.5% min, Thermo Scientific Chemicals

Fertilizers, photography, fabric dyeing, zinc coating, electroplating, and as a separator of Zr and Hf
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 036247.A1
also known as 036247-A1
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
Add to cart
Ammonium thiocyanate, ACS, 97.5% min, Thermo Scientific Chemicals
Catalog number036247.A1
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1762-95-4
IUPAC Nameammonium cyanosulfanide
Molecular FormulaCH4N2S
InChI KeySOIFLUNRINLCBN-UHFFFAOYSA-N
SMILES[NH4+].[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.5% NH₄SCN min
Impurity contentCl 0.005%
Impurity contentIodine-consuming substances 0.004 meq/g
Insoluble material0.005%
Trace Metals(as Pb) 5ppm
View more
Ammonium thiocyanate has many uses in manufacturing chemicals such as metal thiocyanates [CuSCN, Ca(SCN)2, NaSCN, KSCN], and in a number of pharmaceuticals. It is used in freezing solutions, pickling, printing, and corrosion inhibitor against acid gases. It is used in photography industry as a stabilizer or accelerator. Used in fertilizers, fabric dyeing, zinc coating, electroplating, and as a separator of Zr and Hf, also Au and Fe.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only