Thermo Scientific Chemicals

Nickel silicide, 99% (metals basis excluding Co), Co 0.1-1%, Thermo Scientific Chemicals

Catalog number: 036251.18
50 g, Each
Thermo Scientific Chemicals

Nickel silicide, 99% (metals basis excluding Co), Co 0.1-1%, Thermo Scientific Chemicals

Catalog number: 036251.18
50 g, Each
Quantity
Catalog number: 036251.18
also known as 036251-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12059-14-2
IUPAC Name
dinickeliosilane
Molecular Formula
Ni2Si
InChI Key
RUFLMLWJRZAWLJ-UHFFFAOYSA-N
SMILES
[Ni]=[Si]=[Ni]
Total Metal Impurities
1% max.
Impurity content
Co 0.1-1%

Description

Ni silicides have been widely investigated for contact materials in microelectronic devices. Electroplating is another major use of the metal. nickel silicide/silicon nanowire heterostructures have been used to produce field-effect transistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text