Thermo Scientific Chemicals

Ytterbium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 036252.22
100 g, Each
Thermo Scientific Chemicals

Ytterbium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 036252.22
100 g, Each
Quantity
Catalog number: 036252.22
also known as 036252-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-37-0
IUPAC Name
diytterbium(3+) trioxidandiide
Molecular Formula
O3Yb2
InChI Key
UZLYXNNZYFBAQO-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Yb+3].[Yb+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Ytterbium(III) oxide is being applied to numerous fiber amplifier and fiber optic technologies. High purity Ytterbium Oxide are widely applied as a doping agent for garnet crystals in lasers a important colorant in glasses and porcelain enamel glazes.. As Ytterbium Oxide has a significantly higher emissivity in the infrared range a higher radiant intensity is obtained with Ytterbium-based payloads.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text