Thermo Scientific Chemicals

Dihydrogen hexachloroplatinate(IV) hexahydrate, ACS, Premion™, 99.95% (metals basis), Pt 37.5% min, Thermo Scientific Chemicals

Catalog number: 036259.06
5 g, Each
Thermo Scientific Chemicals

Dihydrogen hexachloroplatinate(IV) hexahydrate, ACS, Premion™, 99.95% (metals basis), Pt 37.5% min, Thermo Scientific Chemicals

Catalog number: 036259.06
5 g, Each
Quantity
Catalog number: 036259.06
also known as 036259-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18497-13-7
IUPAC Name
platinum(4+) dihydrogen hexahydrate hexachloride
Molecular Formula
Cl6H14O6Pt
InChI Key
PIJUVEPNGATXOD-UHFFFAOYSA-J
SMILES
[H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
Assay (unspecified)
37.50% Pt min
Comment
Suitability for potassium determinations P.T.
Impurity content
Alkali and other salts (as sulfates) 0.05%
Solution Test
in alcohol P.T.

Description

Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride flowed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text