Thermo Scientific Chemicals

Ethanolamine, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 036260.AP
500 mL, Each
Thermo Scientific Chemicals

Ethanolamine, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 036260.AP
500 mL, Each
Quantity
Catalog number: 036260.AP
also known as 036260-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-43-5
IUPAC Name
2-aminoethan-1-ol
Molecular Formula
C2H7NO
InChI Key
HZAXFHJVJLSVMW-UHFFFAOYSA-N
SMILES
NCCO
Color (APHA)
15
Residual water
0.3%
Impurity content
Fe 5ppm
Trace Metals
(as Pb) 5ppm

Description

It is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text