Ethanolamine, ACS, 99+%, Thermo Scientific Chemicals
Ethanolamine, ACS, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethanolamine, ACS, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 500 mL
500 mL
Catalog number 036260.AY
also known as 036260-AY
Price (USD)/ Each
180.00
-
Add to cart
Quantity:
4 x 500 mL
Request bulk or custom format
Price (USD)/ Each
180.00
Add to cart
Ethanolamine, ACS, 99+%, Thermo Scientific Chemicals
Catalog number036260.AY
Price (USD)/ Each
180.00
-
Add to cart
Chemical Identifiers
IUPAC Name2-aminoethan-1-ol
Molecular FormulaC2H7NO
InChI KeyHZAXFHJVJLSVMW-UHFFFAOYSA-N
SMILESNCCO
Molecular Weight (g/mol)61.08
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)15
Residual water0.3%
Impurity contentFe 5ppm
Trace Metals(as Pb) 5ppm
It is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only