Mercury(II) oxide, yellow, ACS, 99.0% min, Thermo Scientific Chemicals
Mercury(II) oxide, yellow, ACS, 99.0% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercury(II) oxide, yellow, ACS, 99.0% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036261.22
also known as 036261-22
Price (USD)/ Each
98.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
116.00 
Save 17.35 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
98.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
116.00 
Save 17.35 (15%)
Add to cart
Mercury(II) oxide, yellow, ACS, 99.0% min, Thermo Scientific Chemicals
Catalog number036261.22
Price (USD)/ Each
98.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
116.00 
Save 17.35 (15%)
-
Add to cart
Chemical Identifiers
CAS21908-53-2
IUPAC Nameoxomercury
Molecular FormulaHgO
InChI KeyUKWHYYKOEPRTIC-UHFFFAOYSA-N
SMILESO=[Hg]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0% HgO min
CommentResidue after reduction 0.05%
Impurity contentCl 0.025%
Impurity contentFe 0.003%
Impurity contentInsoluble in dilute hydrochloric acid 0.03%
View more
Mercury(II) oxide production of mercury. It is also used as a material for cathodes for mercury batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only