Thermo Scientific Chemicals

Zirconium(IV) oxide, yttria stabilized, 99% (metals basis excluding Hf), Hf 4% max, Thermo Scientific Chemicals

Catalog number: 036272.36
500 g, Each
Thermo Scientific Chemicals

Zirconium(IV) oxide, yttria stabilized, 99% (metals basis excluding Hf), Hf 4% max, Thermo Scientific Chemicals

Catalog number: 036272.36
500 g, Each
Quantity
Catalog number: 036272.36
also known as 036272-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1314-23-4
IUPAC Name
dioxozirconium
Molecular Formula
O2Zr
InChI Key
MCMNRKCIXSYSNV-UHFFFAOYSA-N
SMILES
O=[Zr]=O
Impurity content
Hf 2-4%
Total Metal Impurities excluding Hf
1% max.

Description

Instead of lime for the oxyhydrogen light. As organic intermediate and in pharmaceutical applications. Used as raw material for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text