Thermo Scientific Chemicals

Indium(III) phosphide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 036283.03
1 g, Each
Thermo Scientific Chemicals

Indium(III) phosphide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 036283.03
1 g, Each
Quantity
Catalog number: 036283.03
also known as 036283-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22398-80-7
IUPAC Name
indiganylidynephosphane
Molecular Formula
InP
InChI Key
GPXJNWSHGFTCBW-UHFFFAOYSA-N
SMILES
P#[In]
Total Metal Impurities
0.001% max.

Description

The water soluble InP/ZnS core/shell QDs are a safer alternative to CdSe/ZnS QDs for biological applications, by comparing their toxicity in vitro (cell culture) and in vivo.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text