Mercury(II) sulfate, ACS, 98.0% min, Thermo Scientific Chemicals
Mercury(II) sulfate, ACS, 98.0% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercury(II) sulfate, ACS, 98.0% min, Thermo Scientific Chemicals

For Use As a Primary Battery Electrolyte, Catalyst in Conversion of Acetylene To Acetaldehyde
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036286.14
also known as 036286-14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7783-35-9
IUPAC Namemercury(2+) sulfate
Molecular FormulaHgO4S
InChI KeyDOBUSJIVSSJEDA-UHFFFAOYSA-L
SMILES[Hg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0% HgSO₄ min
CommentResidue after reduction 0.02%
Impurity contentCl 0.003%
Impurity contentFe 0.005%
Impurity contentMercurous mercury (as Hg) 0.15%
View more
Mercury(II) sulfate is used for gold and silver extraction, and to make other mercury compounds. Used as catalyst in the conversion of acetylene to acetaldehyde and used as a electrolyte in primary battery. It is commonly used as a catalyst in oxymercuration-demercuration, a type of electrophilic addition reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only