Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 036291.18
50 g, Each
Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 036291.18
50 g, Each
Quantity
Catalog number: 036291.18
also known as 036291-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-27-2
IUPAC Name
aluminium(3+) nonahydrate trinitrate
Molecular Formula
AlH18N3O18
InChI Key
SWCIQHXIXUMHKA-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Impurity content
Fe 0.002%
Assay (unspecified)
98.0-102.0% Al(NO₃)₃?9H₂O
Impurity content
Ca 0.005%
Impurity content
Mg 0.001%
Impurity content
SO₄ 0.005%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text