Thermo Scientific Chemicals

Acetic anhydride, ACS, 97+%, Thermo Scientific Chemicals

Catalog number: 036292.AE
100 mL, Each
Thermo Scientific Chemicals

Acetic anhydride, ACS, 97+%, Thermo Scientific Chemicals

Catalog number: 036292.AE
100 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 036292.AE
also known as 036292-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-24-7
IUPAC Name
acetyl acetate
Molecular Formula
C4H6O3
InChI Key
WFDIJRYMOXRFFG-UHFFFAOYSA-N
SMILES
CC(=O)OC(C)=O
Form
Liquid
Appearance (Color)
Colorless to very faint yellow
Assay (GC)
≥97.0%
Impurity content
Cl 5ppm, max
Impurity content
Fe 5ppm, max

Description

It is most suited for the conversion of cellulose into cellulose acetate and in the manufacture of aspirin. It acts as a wood preservative and solvent in examination of wood fat, fatty and volatile oils and detection of rosins. In many organic reactions it acts as a dehydrating agent. Also used in the production of isoamyl alcohol and 2-methyl furan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text