Acetic anhydride, ACS, 97+%, Thermo Scientific Chemicals
Acetic anhydride, ACS, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetic anhydride, ACS, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036292.AP
also known as 036292-AP
Price (USD)/ Each
50.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
59.60 
Save 8.95 (15%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
50.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
59.60 
Save 8.95 (15%)
Add to cart
Acetic anhydride, ACS, 97+%, Thermo Scientific Chemicals
Catalog number036292.AP
Price (USD)/ Each
50.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
59.60 
Save 8.95 (15%)
-
Add to cart
Chemical Identifiers
CAS108-24-7
IUPAC Nameacetyl acetate
Molecular FormulaC4H6O3
InChI KeyWFDIJRYMOXRFFG-UHFFFAOYSA-N
SMILESCC(=O)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Colorless to very faint yellow
Assay (GC)≥97.0%
Impurity contentCl 5ppm, max
Impurity contentFe 5ppm, max
View more
It is most suited for the conversion of cellulose into cellulose acetate and in the manufacture of aspirin. It acts as a wood preservative and solvent in examination of wood fat, fatty and volatile oils and detection of rosins. In many organic reactions it acts as a dehydrating agent. Also used in the production of isoamyl alcohol and 2-methyl furan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only