Aluminum sulfate hydrate, ACS, 98.0-102%, Thermo Scientific Chemicals
Aluminum sulfate hydrate, ACS, 98.0-102%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum sulfate hydrate, ACS, 98.0-102%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 036294.A1
also known as 036294-A1
Price (USD)/ Each
99.65
Online exclusive
110.00 
Save 10.35 (9%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
99.65
Online exclusive
110.00 
Save 10.35 (9%)
Add to cart
Aluminum sulfate hydrate, ACS, 98.0-102%, Thermo Scientific Chemicals
Catalog number036294.A1
Price (USD)/ Each
99.65
Online exclusive
110.00 
Save 10.35 (9%)
-
Add to cart
Chemical Identifiers
CAS17927-65-0
IUPAC Namedialuminium(3+) trisulfate
Molecular FormulaAl2O12S3
InChI KeyDIZPMCHEQGEION-UHFFFAOYSA-H
SMILES[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA98.0-102.0% (titration)
Impurity contentCa: ≤0.01%
Appearance (Color)White to pale cream
FormCrystals or crystalline powder
Impurity contentCl: ≤0.005%
View more
Aluminum sulfate is a chemical compound commonly used as a flocculating agent in the purification of drinking water and waste water treatment plants, and also in paper manufacturing. Used as coagulant in decolorization and COD removal of secondary yeast wastewater effluents. It is also plays a significant role in arsenic removal by coagulation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only